3-(4-Methoxyphenyl)-2-propen-1-ol 95%

Supplier: Ambeed
Danger

Synonyms: 3-(4-Methoxyphenyl)prop-2-en-1-ol

A660765-1G A660765-250MG A660765-5G
77005-748EA 248 USD
77005-748 77005-750 77882-511
3-(4-Methoxyphenyl)-2-propen-1-ol 95%
3-(4-Methoxyphenyl)-2-propen-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00182606
CAS Number: 17581-85-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR