2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide 98%

Supplier: Ambeed
Warning

A660708-1G A660708-5G A660708-250MG
77018-390EA 82 USD
77018-390 77018-392 77891-305
2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide 98%
2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide

Precaución: for Research Use Only

Formula: C₃₀H₃₆Br₂N₂O₄
MDL Number: MFCD27923032
CAS Number: 1088205-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR