Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Benzyloxyindole-2-carboxylic acid ethyl ester, Ethyl 5-(benzyloxy)-1H-indole-2-carboxylate
A660525-1G
A660525-250MG
A660525-25G
A660525-5G
77035-516EA
43.5
USD
77035-516
77035-518
77035-520
77035-522
Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate 95%
Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₈H₁₇NO₃ MW: 295.34 g/mol |
MDL Number:
MFCD00022702 CAS Number: 37033-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...