2-((tert-Butoxycarbonyl)amino)-2-(tetrahydro-2H-pyran-4-yl)acetic acid 97%

Supplier: Ambeed
Warning

A660279-5G A660279-1G A660279-250MG
77018-382EA 119 USD
77018-382 77891-302 77891-303
2-((tert-Butoxycarbonyl)amino)-2-(tetrahydro-2H-pyran-4-yl)acetic acid 97%
2-((tert-Butoxycarbonyl)amino)-2-(tetrahydro-2H-pyran-4-yl)acetic acid
Formula: C₁₂H₂₁NO₅
MDL Number: MFCD04114473
CAS Number: 182287-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR