2-Fluoro-1,3-bis-(trifluoromethyl)benzene 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Fluoro-1,3-bis(trifluoromethyl)benzene, 2,6-Bis(trifluoromethyl)fluorobenzene
A660010-100MG
A660010-1G
A660010-250MG
A660010-5G
77005-742EA
24.41
USD
77005-742
77005-744
77005-746
77673-798
2-Fluoro-1,3-bis-(trifluoromethyl)benzene 97%
2-Fluoro-1,3-bis-(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₈H₃F₇ MW: 232.1 g/mol |
MDL Number:
MFCD04972866 CAS Number: 887268-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...