(Fluoromethyl)triphenylphosphonium tetrafluoroborate 97%

Supplier: Ambeed
Warning

A659151-100MG A659151-1G A659151-250MG A659151-5G A659151-100G
77018-372EA 11 USD
77018-372 77018-374 77018-376 77018-378 77891-299
(Fluoromethyl)triphenylphosphonium tetrafluoroborate 97%
(Fluoromethyl)triphenylphosphonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₉H₁₇BF₅P
MDL Number: MFCD22124052
CAS Number: 96385-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR