4-Fluorophenylboronic acid neopentyl glycol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(4-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
A659012-1G
A659012-250MG
A659012-5G
77018-366EA
30
USD
77018-366
77018-368
77018-370
4-Fluorophenylboronic acid neopentyl glycol ester 97%
4-Fluorophenylboronic acid neopentyl glycol ester
Precaución: for Research Use Only
Formula:
C₁₁H₁₄BFO₂ |
MDL Number:
MFCD05663850 CAS Number: 225916-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...