2-Amino-6-methoxypurine 95%

Supplier: Ambeed
Warning

A657900-10G A657900-1G A657900-5G A657900-25G
77005-726EA 31 USD
77005-726 77005-728 77005-730 77882-505
2-Amino-6-methoxypurine 95%
2-Amino-6-methoxypurine

Precaución: for Research Use Only

Formula: C₆H₇N₅O
MDL Number: MFCD00133009
CAS Number: 20535-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR