1-Benzyl-1H-pyrazol-3-amine 98%

Supplier: Ambeed
Warning

A657894-250MG A657894-5G A657894-1G A657894-25G
77035-508EA 19 USD
77035-508 77035-510 77873-059 77873-060
1-Benzyl-1H-pyrazol-3-amine 98%
1-Benzyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃
MDL Number: MFCD03422517
CAS Number: 21377-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR