2-Bromo-5,6-dihydrobenzo[d]thiazol-7(4H)-one 98%

Supplier: Ambeed
Warning

A657212-100MG A657212-1G A657212-250MG
76992-444EA 38.2 USD
76992-444 76992-446 76992-448
2-Bromo-5,6-dihydrobenzo[d]thiazol-7(4H)-one 98%
2-Bromo-5,6-dihydrobenzo[d]thiazol-7(4H)-one

Precaución: for Research Use Only

Formula: C₇H₆BrNOS
MW: 232.09 g/mol
MDL Number: MFCD13250122
CAS Number: 1201633-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR