6,7-Dichloroquinoline-5,8-dione 98%

Supplier: Ambeed
Warning

A657071-100MG A657071-10G A657071-250MG A657071-5G
76992-436EA 76 USD
76992-436 76992-438 76992-440 76992-442
6,7-Dichloroquinoline-5,8-dione 98%
6,7-Dichloroquinoline-5,8-dione

Precaución: for Research Use Only

Formula: C₉H₃Cl₂NO₂
MDL Number: MFCD00512152
CAS Number: 6541-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR