2-(3-Bromophenyl)-1-phenylbenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Bromophenyl)-1-phenyl-1H-benzoimidazole

A656689-1G A656689-5G
77035-504EA 50.93 USD
77035-504 77035-506
2-(3-Bromophenyl)-1-phenylbenzimidazole 98%
2-(3-Bromophenyl)-1-phenylbenzimidazole

Precaución: for Research Use Only

Formula: C₁₉H₁₃BrN₂
MW: 349.23 g/mol
MDL Number: MFCD12911655
CAS Number: 760212-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR