[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-benzyl]-carbamic acid tert-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylcarbamate

A656308-1G A656308-250MG A656308-100MG A656308-5G
77035-500EA 53 USD
77035-500 77035-502 77873-056 77873-057
[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-benzyl]-carbamic acid tert-butyl ester 98%
[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-benzyl]-carbamic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₈H₂₈BNO₄
MW: 333.23 g/mol
MDL Number: MFCD05863917
CAS Number: 832114-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR