1,2,3,5-Tetra-O-acetyl-β-L-ribofuranose 97%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (2R,3S,4S,5S)-5-(Acetoxymethyl)tetrahydrofuran-2,3,4-triyl triacetate

A655908-1G A655908-5G A655908-25G
77018-352EA 22 USD
77018-352 77018-354 77684-548
1,2,3,5-Tetra-O-acetyl-β-L-ribofuranose 97%, Ambeed.Inc
1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose

Precaución: For research use only.

Formula: C₁₃H₁₈O₉
MW: 318.278 g/mol
MDL Number: MFCD00674547
CAS Number: 144490-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR