5-Amino-8-bromoquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 8-Bromoquinolin-5-amine

A655425-1G A655425-250MG A655425-5G
77035-492EA 64.72 USD
77035-492 77035-494 77035-496
5-Amino-8-bromoquinoline 98%
5-Amino-8-bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
MDL Number: MFCD11110428
CAS Number: 116632-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR