(2-Methylthiophen-3-yl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methylthiophene-3-boronic acid

A655398-100MG A655398-1G A655398-250MG A655398-5G
77035-484EA 35.01 USD
77035-484 77035-486 77035-488 77035-490
(2-Methylthiophen-3-yl)boronic acid 98%
(2-Methylthiophen-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₇BO₂S
MW: 141.98 g/mol
MDL Number: MFCD12032042
CAS Number: 177735-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR