1-(Prop-2-yn-1-yl)pyrrolidine 95%

Supplier: Ambeed
Danger

A654630-1G A654630-250MG A654630-5G
77035-480EA 47 USD
77035-480 77035-482 77573-100
1-(Prop-2-yn-1-yl)pyrrolidine 95%
1-(Prop-2-yn-1-yl)pyrrolidine

Precaución: for Research Use Only

Formula: C₇H₁₁N
MDL Number: MFCD03727329
CAS Number: 7223-42-9
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR