Methyl 4-hydroxy-5-nitronicotinate 97%

Supplier: Ambeed
Warning

A654174-100MG A654174-250MG A654174-5G A654174-1G
76992-424EA 20 USD
76992-424 76992-426 76992-428 77891-292
Methyl 4-hydroxy-5-nitronicotinate 97%
Methyl 4-hydroxy-5-nitronicotinate

Precaución: for Research Use Only

Formula: C₇H₆N₂O₅
MDL Number: MFCD14698053
CAS Number: 1214387-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR