(1-Methyl-1H-benzimidazol-2-yl)methylamine 95%

Supplier: Ambeed
Danger

Synonyms: 2-(Aminomethyl)-1-methyl-1H-benzimidazole, (1-Methyl-1H-benzo[d]imidazol-2-yl)methanamine

A653325-10G A653325-1G A653325-5G A653325-100MG
77018-344EA 427.55 USD
77018-344 77018-346 77018-348 77684-543
(1-Methyl-1H-benzimidazol-2-yl)methylamine 95%
(1-Methyl-1H-benzimidazol-2-yl)methylamine

Precaución: For research use only.

Formula: C₉H₁₁N₃
MW: 161.21 g/mol
MDL Number: MFCD00457746
CAS Number: 20028-40-4
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR