(S)-(-)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(S)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene, (S)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
A651894-100MG
A651894-1G
A651894-250MG
A651894-5G
77035-462EA
9.55
USD
77035-462
77035-464
77035-466
77035-468
(S)-(-)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl 98%
(S)-(-)-6,6'-Dibromo-2,2'-bis(methoxymethoxy)-1,1'-binaphthyl
Precaución: for Research Use Only
Formula:
C₂₄H₂₀Br₂O₄ MW: 532.23 g/mol |
MDL Number:
MFCD03788936 CAS Number: 211560-97-3 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...