2-Cyclopropylethanol 95%

Supplier: Ambeed
Danger

A651636-1G A651636-250MG A651636-25G A651636-5G A651636-10G
76992-412EA 31 USD
76992-412 76992-414 77569-234 77614-020 77886-549
2-Cyclopropylethanol 95%
2-Cyclopropylethanol

Precaución: For research use only.

Formula: C₅H₁₀O
MDL Number: MFCD00040762
CAS Number: 2566-44-1
UN: 1987
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR