3,6-Bis(5-bromothiophen-2-yl)-2,5-bis(2-octyldodecyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 95%

Supplier: Ambeed
Warning

A651403-100MG A651403-250MG
77018-336EA 47 USD
77018-336 77018-338
3,6-Bis(5-bromothiophen-2-yl)-2,5-bis(2-octyldodecyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 95%
3,6-Bis(5-bromothiophen-2-yl)-2,5-bis(2-octyldodecyl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione

Precaución: for Research Use Only

Formula: C₅₄H₈₆Br₂N₂O₂S₂
MDL Number: MFCD27923046
CAS Number: 1260685-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR