12,43-Dichloro-1,4(1,4)-dibenzenacyclohexaphane 97% mix

Supplier: Ambeed
Warning

A651331-100G A651331-1G A651331-25G A651331-5G
77035-454EA 234 USD
77035-454 77035-456 77035-458 77035-460
12,43-Dichloro-1,4(1,4)-dibenzenacyclohexaphane 97% mix
12,43-Dichloro-1,4(1,4)-dibenzenacyclohexaphane

Precaución: for Research Use Only

Formula: C₁₆H₁₄Cl₂
MW: 277.18 g/mol
MDL Number: MFCD09038738
CAS Number: 10366-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR