N-(3-((4-Aminobutyl)amino)propyl)acetamide dihydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N1-Acetylspermidine hydrochloride
A649484-100MG
A649484-250MG
A649484-50MG
A649484-1G
76992-406EA
166.57
USD
76992-406
76992-408
76992-410
77684-535
N-(3-((4-Aminobutyl)amino)propyl)acetamide dihydrochloride 95%
N-(3-((4-Aminobutyl)amino)propyl)acetamide dihydrochloride
Precaución: For research use only.
Formula:
C₉H₂₃Cl₂N₃O MW: 260.20 g/mol |
MDL Number:
MFCD00058261 CAS Number: 34450-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...