(2S,6S)-2,6-Di-tert-butyl-2,3,5,6-tetrahydro-1H-imidazo[1,2-a]imidazole 98%

Supplier: Ambeed
Warning

A648738-100MG A648738-1G A648738-250MG
76992-388EA 154.9 USD
76992-388 76992-390 76992-392
(2S,6S)-2,6-Di-tert-butyl-2,3,5,6-tetrahydro-1H-imidazo[1,2-a]imidazole 98%
(2S,6S)-2,6-Di-tert-butyl-2,3,5,6-tetrahydro-1H-imidazo[1,2-a]imidazole

Precaución: for Research Use Only

Formula: C₁₃H₂₅N₃
MW: 223.35 g/mol
MDL Number: MFCD22690898
CAS Number: 877773-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR