1-(3-Chlorophenyl)-1H-1,2,3-triazole-4-carboxylic acid 96%

Supplier: Ambeed
Warning

A648327-100MG A648327-1G A648327-250MG
77035-446EA 29 USD
77035-446 77035-448 77035-450
1-(3-Chlorophenyl)-1H-1,2,3-triazole-4-carboxylic acid 96%
1-(3-Chlorophenyl)-1H-1,2,3-triazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆ClN₃O₂
MDL Number: MFCD10696379
CAS Number: 944901-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR