[3-(4-Chlorophenyl)-5-isoxazolyl]methanol 97%

Supplier: Ambeed
Warning

Synonyms: (3-(4-Chlorophenyl)isoxazol-5-yl)methanol

A648282-100MG A648282-1G A648282-250MG
77018-328EA 53.05 USD
77018-328 77018-330 77018-332
[3-(4-Chlorophenyl)-5-isoxazolyl]methanol 97%
[3-(4-Chlorophenyl)-5-isoxazolyl]methanol

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO₂
MW: 209.6290 g/mol
MDL Number: MFCD01935957
CAS Number: 206055-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR