2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-didecylfluorene 98%

Supplier: Ambeed
Warning

Synonyms: 9,9-Didecylfluorene-2,7-diboronic acid bis(pinacol) ester, 2,2'-(9,9-Didecyl-9H-fluorene-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

A648266-100MG A648266-1G A648266-250MG
76992-382EA 28 USD
76992-382 76992-384 76992-386
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-didecylfluorene 98%
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-didecylfluorene

Precaución: for Research Use Only

Formula: C₄₅H₇₂B₂O₄
MDL Number: MFCD20275090
CAS Number: 711026-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR