6-Bromo-3-chloro-5-fluoropicolinic acid 95%

Supplier: Ambeed
Warning

A647656-100MG A647656-1G A647656-250MG A647656-50MG
77018-320EA 60 USD
77018-320 77018-322 77018-324 77018-326
6-Bromo-3-chloro-5-fluoropicolinic acid 95%
6-Bromo-3-chloro-5-fluoropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₂BrClFNO₂
MW: 254.441 g/mol
MDL Number: MFCD18256604
CAS Number: 514798-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR