6-Nitro-o-toluonitrile 98%

Supplier: Ambeed
Warning

A647286-1G A647286-25G A647286-5G
77035-428EA 17 USD
77035-428 77035-430 77035-432
6-Nitro-o-toluonitrile 98%
6-Nitro-o-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD00007266
CAS Number: 1885-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR