7-(Benzyloxy)-6-methoxyquinolin-4-ol 98%

Supplier: Ambeed
Warning

A647210-100MG A647210-1G A647210-250MG
77035-422EA 15 USD
77035-422 77035-424 77035-426
7-(Benzyloxy)-6-methoxyquinolin-4-ol 98%
7-(Benzyloxy)-6-methoxyquinolin-4-ol

Precaución: for Research Use Only

Formula: C₁₇H₁₅NO₃
MDL Number: MFCD16877538
CAS Number: 849217-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR