tert-Butyl (3-bromo-4-methylphenyl)carbamate 96%

Supplier: Ambeed
Warning

A646279-1G A646279-5G
76992-358EA 16.98 USD
76992-358 76992-360
tert-Butyl (3-bromo-4-methylphenyl)carbamate 96%
tert-Butyl (3-bromo-4-methylphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂
MW: 286.16 g/mol
MDL Number: MFCD12913704
CAS Number: 515813-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR