2-Iodophenyl trifluoromethanesulfonate 98+%

Supplier: Ambeed
Warning

Synonyms: 2-Iodophenyl Triflate, Trifluoromethanesulphonic Acid 2-Iodophenyl Ester

A646229-100MG A646229-1G A646229-250MG A646229-5G
76992-350EA 6 USD
76992-350 76992-352 76992-354 76992-356
2-Iodophenyl trifluoromethanesulfonate 98+%
2-Iodophenyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₇H₄F₃IO₃S
MDL Number: MFCD07784326
CAS Number: 129112-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR