3-Bromo-2-chlorobenzyl Alcohol 95%

Supplier: Ambeed
Warning

A645690-100MG A645690-250MG A645690-1G
76992-344EA 19.1 USD
76992-344 76992-346 77684-529
3-Bromo-2-chlorobenzyl Alcohol 95%
3-Bromo-2-chlorobenzyl Alcohol

Precaución: For research use only.

Formula: C₇H₆BrClO
MW: 221.48 g/mol
MDL Number: MFCD12923279
CAS Number: 1261524-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR