2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazolium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazol-3-ium chloride

A644690-100MG A644690-1G A644690-250MG
76992-330EA 31.83 USD
76992-330 76992-332 76992-334
2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazolium chloride 98%
2-Chloro-1,3-bis(2,6-diisopropylphenyl)-1H-imidazolium chloride

Precaución: for Research Use Only

Formula: C₂₇H₃₆Cl₂N₂
MW: 459.50 g/mol
MDL Number: MFCD23703069
CAS Number: 1228185-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR