3-(Piperidin-1-ylsulfonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-(Piperidin-1-ylsulphonyl)phenyl)boronic acid

A644378-100MG A644378-1G A644378-250MG
76992-324EA 36.08 USD
76992-324 76992-326 76992-328
3-(Piperidin-1-ylsulfonyl)phenylboronic acid 98%
3-(Piperidin-1-ylsulphonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₄S
MW: 269.1300 g/mol
MDL Number: MFCD07783850
CAS Number: 690662-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR