Potassium tert-butyl N-[2-(trifluoroboranuidyl)ethyl]carbamate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Potassium, Potassium 2-(Boc-aminoethyl)trifluoroborate, Potassium (2-((tert-butoxycarbonyl)amino)ethyl)trifluoroborate, trifluoro-[2-[(2-methylpropan-2-yl)oxycarbonylamino]ethyl]boranuide
A644303-100MG
A644303-1G
A644303-250MG
A644303-5G
A644303-25G
A644303-100G
A644303-10G
77035-408EA
19
USD
77035-408
77035-410
77035-412
77035-414
77602-650
77669-764
77669-766
Potassium tert-butyl N-[2-(trifluoroboranuidyl)ethyl]carbamate 96%
Potassium tert-butyl N-[2-(trifluoroboranuidyl)ethyl]carbamate
Precaución: for Research Use Only
Formula:
C₇H₁₄BF₃NO₂·K MW: 251.096 g/mol |
MDL Number:
MFCD10698613 CAS Number: 926280-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...