Methyl 4-(4-hydroxybutyl)benzoate 95%

Supplier: Ambeed
Warning

A644247-25G A644247-5G
77018-308EA 98 USD
77018-308 77891-284
Methyl 4-(4-hydroxybutyl)benzoate 95%
Methyl 4-(4-hydroxybutyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MDL Number: MFCD12547324
CAS Number: 123910-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR