3-Bromo-4-methoxy-1-methyl-1H-indazole 95%

Supplier: Ambeed
Warning

A643965-100MG A643965-1G A643965-250MG
77018-302EA 79 USD
77018-302 77018-304 77018-306
3-Bromo-4-methoxy-1-methyl-1H-indazole 95%
3-Bromo-4-methoxy-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₉BrN₂O
MDL Number: MFCD27923527
CAS Number: 1779786-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR