1-Boc-4-bromo-1H-indazole 95%

Supplier: Ambeed
Warning

A643729-1G A643729-250MG
77035-404EA 44 USD
77035-404 77035-406
1-Boc-4-bromo-1H-indazole 95%
1-Boc-4-bromo-1H-indazole

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O₂
MDL Number: MFCD11110271
CAS Number: 926922-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR