6,7-Dihydro-1H-indazol-4(5H)-one 95%

Supplier: Ambeed
Warning

A642312-1G A642312-250MG
76992-318EA 215.37 USD
76992-318 76992-320
6,7-Dihydro-1H-indazol-4(5H)-one 95%
6,7-Dihydro-1H-indazol-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD09054891
CAS Number: 499206-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR