Triethyl-2-phosphonobutyrate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(diethoxyphosphoryl)butanoate

A642036-1G A642036-25G A642036-5G
76992-312EA 19.1 USD
76992-312 76992-314 76992-316
Triethyl-2-phosphonobutyrate 98%
Triethyl 2-phosphonobutyrate

Precaución: for Research Use Only

Formula: C₁₀H₂₁O₅P
MW: 252.25 g/mol
MDL Number: MFCD00041347
CAS Number: 17145-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR