N-(6-Chloro-3-formylpyridin-2-yl)pivalamide 97%

Supplier: Ambeed
Warning

A641750-10G A641750-250MG A641750-1G
77035-400EA 1799.29 USD
77035-400 77035-402 77669-746
N-(6-Chloro-3-formylpyridin-2-yl)pivalamide 97%
N-(6-Chloro-3-formylpyridin-2-yl)pivalamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₂O₂
MW: 240.68 g/mol
MDL Number: MFCD11846452
CAS Number: 127446-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR