(S)-3-Amino-3-(4-bromophenyl)propanoic acid 97%

Supplier: Ambeed
Warning

A641315-1G A641315-250MG A641315-5G
77005-688EA 37 USD
77005-688 77005-690 77602-632
(S)-3-Amino-3-(4-bromophenyl)propanoic acid 97%
(S)-3-Amino-3-(4-bromophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MDL Number: MFCD03840456
CAS Number: 275826-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR