Poly(hexamethylene biguanide) hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Arlagard E, Poly(hexamethylene biguanide)hydrochloride, Poly (hexamethylene biguanide) hydrochloride, Polihexanide, Poly(iminocarbonimidoyliminocarbonimidoylimino-1,6-hexanediyl) hydrochloride, Acticide SR 1296, BG 1

A641246-1000G A641246-100G A641246-10G A641246-1G A641246-25G A641246-500G A641246-5G
77005-674EA 128.37 USD
77005-674 77005-676 77005-678 77005-680 77005-682 77005-684 77005-686
Poly(hexamethylene biguanide) hydrochloride 98%
Poly(hexamethylene biguanide) hydrochloride

Precaución: for Research Use Only

Formula: (C₈H₁₇N₅)n·xHCl
MW: (433.038)n g/mol
MDL Number: MFCD00217750
CAS Number: 32289-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR