Ethyl-4-bromo-3-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-bromo-3-nitrobenzoate

A640994-100MG A640994-1G A640994-250MG A640994-5G
76992-306EA 5.31 USD
76992-306 76992-308 76992-310 77613-988
Ethyl-4-bromo-3-nitrobenzoate 97%
Ethyl-4-bromo-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrNO₄
MW: 274.07 g/mol
MDL Number: MFCD09972120
CAS Number: 168473-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR