8-(Benzyloxy)-2-methylimidazo[1,2-a]pyridine 95%

Supplier: Ambeed
Warning

A640833-100MG A640833-1G A640833-250MG
76992-300EA 62.6 USD
76992-300 76992-302 76992-304
8-(Benzyloxy)-2-methylimidazo[1,2-a]pyridine 95%
8-(Benzyloxy)-2-methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O
MW: 238.28 g/mol
MDL Number: MFCD18432284
CAS Number: 79707-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR