(R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: (R)-3-(4-Bromophenyl)-3-((tert-butoxycarbonyl)amino)propanoic acid

A640254-100MG A640254-1G A640254-250MG A640254-5G
77005-656EA 32.89 USD
77005-656 77005-658 77005-660 77005-662
(R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid 97%
(R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₄
MW: 344.20 g/mol
MDL Number: MFCD01320860
CAS Number: 261380-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR