1-Bromo-2,4,5-trifluoro-3-methylbenzene 95%

Supplier: Ambeed
Danger

A639741-1G A639741-250MG A639741-5G
76992-292EA 21.22 USD
76992-292 76992-294 76992-296 76992-298
1-Bromo-2,4,5-trifluoro-3-methylbenzene 95%
1-Bromo-2,4,5-trifluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₄BrF₃
MDL Number: MFCD16621735
CAS Number: 112822-82-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR