Methyl 2,4-dimethyl-5-nitrobenzoate 98%

Supplier: Ambeed
Warning

A639391-1G A639391-5G
77018-274EA 41 USD
77018-274 77018-276
Methyl 2,4-dimethyl-5-nitrobenzoate 98%
Methyl 2,4-dimethyl-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MDL Number: MFCD00184976
CAS Number: 202264-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR